Anti-CAMP Rabbit Polyclonal Antibody
Catalog # ENZOADI915053100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Cathelicidin
- Antigen symbol:CAMP
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Multiple
- Seq:02
- Form:Neat.
- Antigen synonyms:FALL-39|CAP18|CAP-18|LL37|CRAMP|Cathelicidin antimicrobial peptide|18 kDa cationic antimicrobial protein|HSD26|FALL39
- Pk:100 µl
Specifications
About this item
Cyclic AMP (cAMP) is one of the most important second messenger involved as a modulator of physiological processes. A number of hormones are known to activate cAMP through the action of the enzyme Adenylate cyclase which converts ATP to cAMP. cAMP has been shown to be involved in the cardiovascular and nervous systems, immune mechanisms, cell growth and differentiation, and general metabolism.
- For ELISA, Flow Cytometry
Species independent.
Type: Primary
Antigen: CAMP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan