Order Entry
France
ContactUsLinkComponent
gDNA Removal kit
gDNA Removal kit
Supplier:  ENZO LIFE SCIENCES
Certificates
gDNA Removal kit
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Method / Format:
    Enzymatic extraction
  • Prep size:
    Scalable
  • Refine target molecule:
    Total RNA
  • Sample type:
    RNA preps
  • Target molecule:
    RNA
  • Description:
    gDNA removal kit

Specifications

Frequently Bought Together