Order Entry
France
ContactUsLinkComponent
 
Pyruvate assay kit
Catalog # ABNOKA1674
Supplier:  Abnova
CAS Number:  
undefined
Pyruvate assay kit
Catalog # ABNOKA1674
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Pyruvate Assay
  • Sample size:
    10 µl
  • Storage temperature:
    −20 °C
  • Tests per kit:
    100 assays

Specifications

About this item

Pyruvate Assay Kit is for quantitative colorimetric/fluorimetric pyruvate determination.

  • Detection method: Colorimetric, fluorometric
  • Calibration range: Colorimetric assay: 2 to 500 μM; fluorimetric assay: 0,2 to 50 μM
Delivery: The kit includes enzyme mix (10 ml), dye reagent (120 µl) and standard 25 mM pyruvate (400 µl).
Caution: For research use only.