Labels, chlorinated waste
Hazard labels, Paper, Chlorinated waste, Permanent acrylic, Chrome, 150×100 mm
Catalog # BKRS909-401
Supplier: BAKER SELF ADHESIVES LABEL
Jump to:
Product Details & Documents
These chlorinated waste labels have a fluorescent chrome coated woodfree paper face with a permanent acrylic based adhesive.
- Good UV resistant
- Maximum visual impact due to its background colour
- Temperature range: –40 to +100 °C
It offers good adhesion on a wide variety of surfaces including apolar substrates like PE and PP.
: Complies with ISEGA, FDA 175.105 and with BgVV XIV recommendations for direct contact with dry non-fatty foodstuff.
Specifications
- Colour:Chrome
- Description:Hazard labels
- Dimensions:150×100 mm
- Material:Paper
- Legend:Chlorinated waste
- Adhesive:Permanent acrylic



