Order Entry
France
Orders LinkContactUsLinkComponent
 

Hexyl nicotinate 98%

Irritant
Hexyl nicotinate 98%
Certificates
Hexyl nicotinate 98%

2 Options Available Below

Product Details & Documents
Synonyms:  Nicotinic Acid Hexyl Ester
  • CAS Number:23597-82-2
  • Formula:C₁₂H₁₇NO₂
  • MDL Number:MFCD00023587
  • Molecular Weight:207.27 g/mol
  • Boiling Pt:329 °C (760 mmHg)
  • Density:1.02 g/cm³
  • Flash Pt:128 °C
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4890 to 1.4910 (20°C, 589 nm)
  • Water:
    ≤0.3 %
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 46,80 €
    46,80 €
    1 * 5 g
       46,80 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 149,00 €
    149,00 €
    1 * 25 g
       149,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.