Methyl docosanoate 99% analytical standard for GC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:929-77-1
:
Formula:C₂₃H₄₆O₂
MDL Number:MFCD00009347
:354.62 g/mol
Melting Pt:54…56 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Flakes
- Infrared spectrum:Conforms
- Melting point:53 to 56 °C
- Solubility:(5 % in Hexane) Clear colorless solution
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together