Order Entry
France
Orders LinkContactUsLinkComponent
Drying trays
Drying trays
Drying trays
Drying trays
Made of acid resistant stainless steel.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning
Product Family Options
  • W×D×H
  • 180×130×40 mm
    1 * 1 Pce
    1 * 1 Pce - 182,00 €
    182,00 €
    1 * 1 Pce
       182,00 €
  • 300×200×40 mm
    1 * 1 Pce
    1 * 1 Pce - 207,00 €
    207,00 €
    1 * 1 Pce
       207,00 €
  • 400×300×40 mm
    1 * 1 Pce
    1 * 1 Pce - 230,00 €
    230,00 €
    1 * 1 Pce
       230,00 €
  • 500×300×40 mm
    1 * 1 Pce
    1 * 1 Pce - 232,00 €
    232,00 €
    1 * 1 Pce
       232,00 €