VWR® Protection Covers
Supplier: VWR Collection
Certificates
About this item
Manufactured from a specially formulated breathable microporous fabric, these covers are distinguished by their superior barrier protection capabilities.
- Superior fluid and particulate barrier
- Lowest level of particle shedding
- Light chemical splash resistance, and maximum tear strength
- Tunnelled elastic, along the bottom perimeter, ensures a secure fit
- Serged seams
VWR® cleanroom protection covers are suitable for GMP/ISO cleanrooms of life science and other critical manufacturing sites. They are customised to fit machines, equipment, and carts in the production area. These covers provide barrier protection against fluids, biologicals, particulates, and light chemical splashes.
Bacterial Filtration Efficiency: 99,9%
Level I per Helmke Drum test
Certifications: Covers are rigorously tested, and manufactured in an ISO certified facility under stringent process controls, to ensure that each item meets exacting quality standards, and performs to specification.
Lot-controlled and validated through independent lab testing.
Lot-controlled and validated through independent lab testing.