Lithium isotopic standard solution, for ICP, CRM, VHG
Catalog # PROOVHG-LIS6LIZ-10
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Concentration:100 mg/L
- Matrix:2% HNO₃
- Pk:100 mL
- Element:Lithium
- Symbol:Li
Specifications
About this item
This solution is intended for use as a calibration or reference standard for inductively coupled plasma mass spectrometry (ICP-MS) where known values for isotopic abundance are required.
Certifications: Traceable to NIST, SRM.