Standards for water pollution QC check, sample hardness, VHG
Référence PROOVHG-QWSHRD-250
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:250 mL
- Description:Water analysis standard for hardness
- Type:Reference standard
Specifications
About this item
This product is intended for use as a quality control check for methods related to hardness measurements includes calcium, magnesium, sodium, calcium hardness and total hardness as CaCO₃.
This standard has been prepared as a whole volume standard and is ready to be analysed as received. This standard is not preserved.
Certifications: Traceable to NIST SRM.