Esomeprazole, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1249789.
- 1249803.
- 1249790.
- Description:
- Esomeprazole Magnesium
- Esomeprazole Strontium
- Esomeprazole Sodium
- CAS No.:
- 217087-09-7
- 934714-36-0
- 161796-78-7
- Formula:
- C₃₄H₄₂MgN₆O₉S₂
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1249789.Esomeprazole MagnesiumSpecifications:
More
Specifications
Cat. No.1249789.DescriptionEsomeprazole MagnesiumCAS No.217087-09-7FormulaC₃₄H₄₂MgN₆O₉S₂ - 1249803.Esomeprazole StrontiumSpecifications:
More
Specifications
Cat. No.1249803.DescriptionEsomeprazole StrontiumCAS No.934714-36-0Formula - 1249790.Esomeprazole SodiumSpecifications:
More
Specifications
Cat. No.1249790.DescriptionEsomeprazole SodiumCAS No.161796-78-7Formula
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



