Order Entry
France
ContactUsLinkComponent
Organic reference standard, Sulfamethoxazole, Ph.Eur., RS
Organic reference standard, Sulfamethoxazole, Ph.Eur., RS
Catalog # EDQMS2100000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Sulfamethoxazole, Ph.Eur., RS
Catalog # EDQMS2100000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    723-46-6
  • Description:
    Sulfamethoxazole
  • Formula:
    C₁₀H₁₁N₃O₃S
  • Storage temperature:
    Ambient
  • Description:
    Sulfamethoxazole

Specifications

About this item

  • In Monograph 0108
  • Pharmaceutical applications