Order Entry
France
ContactUsLinkComponent
Human acute myeloid leukemia (AML) plasma
Human acute myeloid leukemia (AML) plasma
Catalog # PB006C-1-AML
CAS Number:  
Human acute myeloid leukemia (AML) plasma
Catalog # PB006C-1-AML
CAS Number:  

Specifications

  • Pk:
    2 mL
  • Description:
    Human acute myeloid leukemia (AML) plasma, cryopreserved

Specifications

About this item

Plasma used for researching the causes, diagnosis, supportive care and treatment of acute myeloid leukemia (AML). Collected from IRB-consented volunteer donors blood diagnosed with acute myeloid leukemia (AML).

No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.

Storage: Store at −80 °C.

Caution: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.