Order Entry
France
ContactUsLinkComponent
Human plasma, rheumatoid arthritis (RA)
Human plasma, rheumatoid arthritis (RA)
Catalog # PB006C-1-RA
CAS Number:  
Human plasma, rheumatoid arthritis (RA)
Catalog # PB006C-1-RA
CAS Number:  

Specifications

  • Pk:
    2 mL
  • Description:
    Plasma, rheumatoid arthritis (RA), cryopreserved

Specifications

About this item

Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of rheumatoid arthritis (RA). Collected from IRB-consented donors diagnosed with RA.

No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.

Storage: Store at −80 °C.

Caution: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.