Order Entry
France
ContactUsLinkComponent
Human plasma, hepatitis C virus (HCV)
Human plasma, hepatitis C virus (HCV)
Catalog # PB006C-1-HCV
CAS Number:  
Human plasma, hepatitis C virus (HCV)
Catalog # PB006C-1-HCV
CAS Number:  

Specifications

  • Pk:
    2 mL
  • Description:
    Plasma, hepatitis C virus (HCV), cryopreserved

Specifications

About this item

Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of hepatitis C virus (HCV). Collected from IRB-consented donors diagnosed with HCV.

Storage: Store at −80 °C.

Caution: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.