Dacarbazine, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1162308.
- 1162320.
- 1162330.
- Description:
- Dacarbazine
- Dacarbazine Related Compound A
- Dacarbazine Related Compound B
- Synonym(s):
- 5-aminoimidazole-4-carboxamide hydrochloride
- 2-Azahypoxanthine monohydrate
- CAS No.:
- 4342-03-4
- 72-40-2
- 7151-03-3
- Formula:
- C₆H₁₀N₆O
- C₄H₇ClN₄O
- C₄H₃N₅O
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1162308.DacarbazineSpecifications:
More
Specifications
Cat. No.1162308.DescriptionDacarbazineSynonym(s)CAS No.4342-03-4FormulaC₆H₁₀N₆O - 1162320.Dacarbazine Related Compound ADacarbazine Related Compound AUSP reference standardVial50 mgLog in for availability1 * 50 mg1 210,00 €Specifications:
More
Specifications
Cat. No.1162320.DescriptionDacarbazine Related Compound ASynonym(s)5-aminoimidazole-4-carboxamide hydrochlorideCAS No.72-40-2FormulaC₄H₇ClN₄O - 1162330.Dacarbazine Related Compound BDacarbazine Related Compound BUSP reference standardVial50 mgLog in for availability1 * 50 mg1 510,00 €Specifications:
More
Specifications
Cat. No.1162330.DescriptionDacarbazine Related Compound BSynonym(s)2-Azahypoxanthine monohydrateCAS No.7151-03-3FormulaC₄H₃N₅O



