Cortisone, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1150003.
- 1149999.
- Description:
- Cortisone Acetate
- Cortisone
- Synonym(s):
- 17, 21-Dihydroxypregna-4-ene-3,11,20-trione
- CAS No.:
- 50-04-4
- 53-06-5
- Formula:
- C₂₃H₃₀O₆
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1150003.Cortisone AcetateSpecifications:
More
Specifications
Cat. No.1150003.DescriptionCortisone AcetateSynonym(s)CAS No.50-04-4FormulaC₂₃H₃₀O₆ - 1149999.CortisoneSpecifications:
More
Specifications
Cat. No.1149999.DescriptionCortisoneSynonym(s)17, 21-Dihydroxypregna-4-ene-3,11,20-trioneCAS No.53-06-5Formula



