Order Entry
France
Orders LinkContactUsLinkComponent
Loratadine, Reference Standards

Loratadine, Reference Standards

Supplier:   USP
Loratadine, Reference Standards
Supplier:  USP
Certificats

4 Options disponibles ci-dessous

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Spécifications de la famille de produits

Spécifications

Spécifications

  • Catalog No:
  • 1370270.
  • 1370280.
  • 1370291.
  • 1370269.
  • Description:
  • Loratadine
  • Loratadine Related Compound A
  • Loratadine Related Compound B
  • Loratadine Related Compound C
  • Synonym(s):
  • 8-Chloro-5,6-dihydro-11-(piperidin-4-ylidene)-11H-benzo[5,6]cyclohepta[1,2-b]pyridine
  • 8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
  • 8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one
  • CAS No.:
  • 79794-75-5
  • 100643-71-8
  • 38092-89-6
  • 31251-41-9
  • Formula:
  • C₂₂H₂₃ClN₂O₂
  • C₁₉H₁₉ClN₂
  • C₂₀H₂₁ClN₂
  • C₁₄H₁₀ClNO
  • UN No.:
  • 3077

Spécifications

Product Family Options

Informations sur le produit

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1370270.
    Loratadine
    Loratadine
    USP reference standard
    Vial
    200 mg
    Connexion pour la disponibilité
    Connexion pour la disponibilité
    1 * 200 mg
    1 * 200 mg - 646,00 €
    646,00 €
    1 * 200 mg
       646,00 €

    Spécifications :
    More
    Specifications
    Cat. No.
    1370270.
    Description
    Loratadine
    Synonym(s)
    CAS No.
    79794-75-5
    Formula
    C₂₂H₂₃ClN₂O₂
    UN No.
    3077
  • 1370280.
    Loratadine Related Compound A
    Loratadine Related Compound A
    USP reference standard
    Vial
    15 mg
    Connexion pour la disponibilité
    Connexion pour la disponibilité
    1 * 15 mg
    1 * 15 mg - 1 170,00 €
    1 170,00 €
    1 * 15 mg
       1 170,00 €

    Spécifications :
    More
    Specifications
    Cat. No.
    1370280.
    Description
    Loratadine Related Compound A
    Synonym(s)
    8-Chloro-5,6-dihydro-11-(piperidin-4-ylidene)-11H-benzo[5,6]cyclohepta[1,2-b]pyridine
    CAS No.
    100643-71-8
    Formula
    C₁₉H₁₉ClN₂
    UN No.
  • 1370291.
    Loratadine Related Compound B
    Loratadine Related Compound B
    USP reference standard
    Vial
    15 mg
    Connexion pour la disponibilité
    Connexion pour la disponibilité
    1 * 15 mg
    1 * 15 mg - 1 390,00 €
    1 390,00 €
    1 * 15 mg
       1 390,00 €

    Spécifications :
    More
    Specifications
    Cat. No.
    1370291.
    Description
    Loratadine Related Compound B
    Synonym(s)
    8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
    CAS No.
    38092-89-6
    Formula
    C₂₀H₂₁ClN₂
    UN No.
  • 1370269.
    Loratadine Related Compound C
    Loratadine Related Compound C
    USP reference standard
    Vial
    15 mg
    Connexion pour la disponibilité
    Connexion pour la disponibilité
    1 * 15 mg
    1 * 15 mg - 1 180,00 €
    1 180,00 €
    1 * 15 mg
       1 180,00 €

    Spécifications :
    More
    Specifications
    Cat. No.
    1370269.
    Description
    Loratadine Related Compound C
    Synonym(s)
    8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one
    CAS No.
    31251-41-9
    Formula
    C₁₄H₁₀ClNO
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.