Order Entry
France
Orders LinkContactUsLinkComponent
Fluorometholone, Reference Standards

Fluorometholone, Reference Standards

Supplier:   USP
Fluorometholone, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1278007.
  • 1278109.
  • 1278018.
  • Description:
  • Fluorometholone
  • Fluorometholone Acetate
  • Fluorometholone Related Compound A
  • Synonym(s):
  • 11β,17α-Dihydroxy-6α-methylpregna-1,4-diene-3,20-dione
  • CAS No.:
  • 426-13-1
  • 3801-06-7
  • 6870-94-6
  • Formula:
  • C₂₂H₂₉FO₄
  • C₂₄H₃₁FO₅
  • C₂₂H₃₀O₄

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1278007.
    Fluorometholone
    Fluorometholone
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 405,00 €
    405,00 €
    1 * 200 mg
       405,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1278007.
    Description
    Fluorometholone
    Synonym(s)
    CAS No.
    426-13-1
    Formula
    C₂₂H₂₉FO₄
  • 1278109.
    Fluorometholone Acetate
    Fluorometholone Acetate
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 400,00 €
    400,00 €
    1 * 200 mg
       400,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1278109.
    Description
    Fluorometholone Acetate
    Synonym(s)
    CAS No.
    3801-06-7
    Formula
    C₂₄H₃₁FO₅
  • 1278018.
    Fluorometholone Related Compound A
    Fluorometholone Related Compound A
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 270,00 €
    1 270,00 €
    1 * 15 mg
       1 270,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1278018.
    Description
    Fluorometholone Related Compound A
    Synonym(s)
    11β,17α-Dihydroxy-6α-methylpregna-1,4-diene-3,20-dione
    CAS No.
    6870-94-6
    Formula
    C₂₂H₃₀O₄