Order Entry
France
ContactUsLinkComponent
Human Recombinant CD137 (from CHO cells)
Human Recombinant CD137 (from CHO cells)
Catalog # PRSI90-409
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant CD137 (from CHO cells)
Catalog # PRSI90-409
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Biological activity:
    Measured by its binding ability in a functional ELISA.
  • Protein synonyms:
    CD137 Antigen|Tumor Necrosis Factor Receptor Superfamily Member 9|T Cell Antigen ILA|TNFRSF9|MGC2172|ILA|CDw137|4-1BB|4-1BB Ligand Receptor T Cell|Antigen 4-1BB Homolog
  • Protein/peptide name:
    CD137
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    19-184
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.

Specifications

About this item

Human CD137 (4-1BB) is a costimulatory molecule of the tumour necrosis factor (TNF) receptor superfamily. The glycoprotein 4-1BB is expressed mainly on activated CD4 + and CD8 + T cells and binds to a high-affinity ligand (4-1BBL) expressed on several antigen-presenting cells such as macrophages and activated B cells. Upon ligand binding, 4-1BB is associated with the tumour receptor-associated factors (TRAF), the adaptor protein and mediates downstream signalling events including the activation of NF-kappaB and cytokine production. 4-1BB signalling either by binding to 4-1BBL or by antibody ligation delivers signals for T cell activation and growth as well as monocyte proliferation and B cell survival, and plays a important role in the amplification of T cell-mediated immune responses. In contrast, it can also enhance activation-induced T cell apoptosis when triggered by engagement of the TCR/CD3 complex. In addition, the 4-1BB/4-1BBL costimulatory pathway has been shown to augment secondary CTL responses to several viruses and increase antitumour immunity. 4-1BB is therefore a promising candidate for immunotherapy of human cancer.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.