Order Entry
France
Orders LinkContactUsLinkComponent
Mouse recombinant Granzyme D

Mouse recombinant Granzyme D

50 µG
Catalog # PRSI92-677
Supplier:   ProSci Inc.
Mouse recombinant Granzyme D
Catalog # PRSI92-677
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant Granzyme D

50 µG

CAS Number:  
Price
724,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Granzyme D is a member of the granzyme family of the serine proteases which plays a role in the induction of apoptosis. T cells, lymphohematopoietic stromal cells, and granulated metrial gland cells express granzyme D, but the function of granzyme D is unknown. Previous studies reported that granzyme D is developmentally regulated during pregnancy together with granzymes E, F, and G in granulated metrial gland cells and is upregulated by IL-2 and IL-15. Granzyme D was also suggested to have a role in stromal cell-lymphocyte interactions.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Documents

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    PRSI92-677
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Granzyme D
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM HEPES, 150mM NaCl, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Protein synonyms:
    Granzyme D|Gzmd
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Tag sequence:
    C-6 His tag
  • Sequence:
    Ile21-Leu252
  • Tested applications:
    Bioassay
  • Species:
    Mouse
  • Molecular weight:
    26.8 kD

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.