Order Entry
France
ContactUsLinkComponent
Mouse recombinant SLAMF5
Mouse recombinant SLAMF5
Catalog # PRSI92-566
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant SLAMF5
Catalog # PRSI92-566
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
  • Species:
    Mouse
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Ly-9B|SLAM family member 5|Leukocyte differentiation antigen CD84|CD84 molecule|CD84 antigen|Cd84|SLAMF5|Signaling lymphocytic activation molecule 5
  • Protein/peptide name:
    SLAMF5
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    23.8 kD
  • Sequence:
    Lys22-Pro223
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

CD84, also called SLAMF5, is a member of the CD2 subgroup of the immunoglobulin receptor superfamily. Members of this CD2 subgroup mediate signal transduction through the interaction of its immunoreceptor tyrosine-based switch motifs (ITSM) in the intracellular region and the SH2 domain of adaptor molecules SAP (SLAM-associated protein) and EAT-2 (EWS-activated transcript 2), and accordingly modulate both adaptive and innate immune responses. CD84 expression has been documented on several hematopoietic cell types, including monocytes, macrophages, dendritic cells, B lymphocytes, and platelets. Activation of cell surface CD84 initiates a signallling cascade involving its intra-cytoplasmic tyrosine residues that results in Bcl-2 upregulation, which in turn enhances cell survival. Either immunoneutralisation or blockade of CD84 with a CD84 extracellular domain protein fragment induces cell death in vitro and in vivo.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.