Order Entry
France
ContactUsLinkComponent
Mouse recombinant NGF beta (from E. coli)
Mouse recombinant NGF beta (from E. coli)
Catalog # PRSI92-518
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant NGF beta (from E. coli)
Catalog # PRSI92-518
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Environmentally Preferable:
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Beta-nerve growth factor/ Beta-NGF/ Ngf
  • Protein/peptide name:
    NGF beta
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    13.5 kD
  • Sequence:
    Ser122-Gly241
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM Tris,150mM NaCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Mouse beta -NGF is a neurotrophic factor structurally related to BDNF, NT-3 and NT-4. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. beta -NGF is a potent neurotrophic factor that signals through its receptor beta -NGFR, and plays a crucial role in the development and preservation of the sensory and sympathetic nervous systems. beta -NGF also acts as a growth and differentiation factor for B lymphocytes, and enhances B-cell survival.

  • This recombinant protein can be used for biological assays. For research use only.

This recombinant protein can be used for biological assays. For research use only.