Order Entry
France
ContactUsLinkComponent
Human recombinant NECAP2 (from E. coli)
Human recombinant NECAP2 (from E. coli)
Catalog # PRSI92-195
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant NECAP2 (from E. coli)
Catalog # PRSI92-195
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    N, C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Adaptin ear-binding coat-associated protein 2|NECAP2|NECAP endocytosis associated protein 2
  • Protein/peptide name:
    NECAP2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    31.5 kD
  • Sequence:
    Met1-Phe263
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

NECAP2 belongs to the NECAP family. The WXXF motifs mediate binding of accessory proteins to the ear-domain of AP-1, GGAs and AP-2 through hydrophobic interactions. Adaptin ear-binding coat-associated protein 2 can interacts with AP1G1 and AP2A1 components of the adapter protein complex AP-1 and AP-2. It also interacts with the GAE domain proteins GGA1, GGA2 and GGA3.

Fusion-Tag: N, C-6 His tag

This recombinant protein can be used for biological assays. For research use only.