Order Entry
France
ContactUsLinkComponent
Human recombinant lymphocyte Antigen 6H (from cells)
Human recombinant lymphocyte Antigen 6H (from cells)
Catalog # PRSI91-765
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant lymphocyte Antigen 6H (from cells)
Catalog # PRSI91-765
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Ly-6H|LY6H|Lymphocyte Antigen 6H
  • Protein/peptide name:
    Lymphocyte Antigen 6H
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    10.9 kD
  • Sequence:
    Leu26-Gly115
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Lymphocyte Antigen 6H (LY6H) is a novel member of the LY6 family of glycosylphosphatidylinositol-anchored cell surface glycoproteins. LY6H contains one UPAR/Ly6 domain. Human LY6H is synthesized as a 140 amino acid precursor that contains a 25 amino acid signal sequence, 20 amino acid propeptide that is removed in the mature form, and a 90 amino acid mature chain. LY6H is highly expressed in the brain (cerebral cortex, amygdala, hippocampus and subthalamic nucleus) and in acute human leukemic cell line MOLT-3. It is also found in lower levels in testis, pancreas, small intestine and colon. It has been shown that LY6H may play a role in both the central nervous system and the immune system.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag