Order Entry
France
ContactUsLinkComponent
Human recombinant LILRA2 (from cells)
Human recombinant LILRA2 (from cells)
Catalog # PRSI91-754
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant LILRA2 (from cells)
Catalog # PRSI91-754
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    ILT-1|Immunoglobulin-like Transcript 1|Leukocyte Immunoglobulin-like Receptor Subfamily A Member 2|Leukocyte Immunoglobulin-like Receptor 7|LIR-7|CD85h|ILT1|LIR7|LILRA2|CD85 Antigen-like Family Member H
  • Protein/peptide name:
    LILRA2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    44.8 kD
  • Sequence:
    Gly24-Ser420
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Leukocyte Immunoglobulin-Like Receptor Subfamily A Member 2 (LILRA2) is a single-pass type I membrane protein. LILRA2 is expressed predominantly on monocytes and B cells, and at lower levels on dendritic cells and natural killer cells. LILRA2 contains four Ig-like C2-type domains, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with transmembrane regions containing a charged arginine residue, may initiate stimulatory cascades. LILRA2 does not bind class I MHC antigens.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag