Human recombinant CEACAM7 (from cells)
Catalog # PRSI91-684
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Environmentally Preferable:
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:CEACAM7|Carcinoembryonic Antigen-Related Cell Adhesion Molecule 7|Carcinoembryonic Antigen CGM2|CGM2
- Protein/peptide name:CEACAM7
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:13.1 kD
- Sequence:Thr36-Phe142
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Carcinoembryonic Antigen-Related Cell Adhesion Molecule 7 (CEACAM7) is a member of the immunoglobulin superfamily A and CEA family. CEACAM7 localizes to the cell membrane and contains one Ig-like C2-type domain and one Ig-like V-type domain. The expression of CEACAM7 is significantly decreased in rectal cancer. Differences in CEACAM7 expression levels between long-term survivors and those with recurrent disease introduce a potential tumor marker to define a subset of patients who benefit most from adjuvant therapy.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag