Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:EPH-like Kinase 11|EHK3|EK11|EPH Homology Kinase 3|EPHA7|EHK-3|hEK11|Ephrin Type-A Receptor 7
- Protein/peptide name:Ephrin Type-A Receptor 7
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:60.19 kD
- Sequence:Gln28-Ile556
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Ephrin Type-A Receptor 7 (EPHA7) is a single-pass type I membrane protein which belongs to the Eph family of transmembrane receptor tyrosine kinases. It contains two fibronectin type-III domains, one protein kinase domain and one SAM (sterile alpha motif) domain. EPHA7 is a receptor for members of the ephrin-A family. Eph receptors are largely expressed throughout the ectoderm, mesoderm, and endoderm of vertebrate embryos. EPHA7 functions as a repulsive guidance molecule during the targeting of retinal axons to the superior colliculus and of neocortical axons to the thalamus. EPHA7 is expressed at a substantial level in most human lung cancers. The high expression of EPHA7 protein may participate in the malignancy transformation, invasion progression and metastasis of primary hepatocellular carcinoma. EPHA7 may involve in smoking related lung carcinogenesis.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag