Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Environmentally Preferable:
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:NLGN1|KIAA1070|Neuroligin-1
- Protein/peptide name:Neuroligin 1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:71.5 kD
- Sequence:Gln46-Leu676
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Neuroligin-1 is a single-pass type I transmembrane protein which belongs to the type-B Carboxylesterase/Lipase family. Neuroligins are cell-adhesion molecules located at the postsynaptic side of the synapse. Neuroligins interact with beta-neurexins and this interaction is involved in the formation of functional synapses. Neurexins and Neuroligins are cell adhesion molecules present in excitatory and inhibitory synapses, and they are required for correct neuron network function. These proteins are found at the presynaptic and postsynaptic membranes. Neuroligin-1 is a neuronal cell surface protein which is thought to be involved in cell-cell-interactions by forming intercellular junctions through binding to beta-neurexins. It seems to play role in formation or maintenance of synaptic junctions. It triggers the de novo formation of presynaptic structures and may be involved in specification of excitatory synapses.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag