Order Entry
France
ContactUsLinkComponent
Human recombinant TREML1 (from cells)
Human recombinant TREML1 (from cells)
Catalog # PRSI91-358
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant TREML1 (from cells)
Catalog # PRSI91-358
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Trem-like Transcript 1 Protein|TREML1|TLT1|Triggering Receptor Expressed on Myeloid Cells-like Protein 1|TLT-1
  • Protein/peptide name:
    TREML1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    16.88 kD
  • Sequence:
    Gln16-Pro162
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

Specifications

About this item

Triggering Receptor Expressed on Myeloid Cells-Like Protein 1 (TREML1) is a single-pass type I membrane protein. TREML1 precursor contains a 15 amino acid signal peptide, a 147 amino acid extracellular domain with an Ig-like V-type (immunoglobulin-like) domain, and 128 amino acid cytoplasmic domain. It can be expressed exclusively in platelets and megakaryocytes (MKs). It is a cell surface receptor that may play a role in the innate and adaptive immune response. TREML1 Sequestered in cytoplasmic vesicles in resting platelets. TREML1 be transported to the cell surface after stimulation by thrombin. Soluble fragments can be released into the serum by proteolysis. The phosphorylated TREML1 can interact with PTPN6 and PTPN11. TREML1 may participate in maintaining vascular hemostasis and regulating coagulation and inflammation at sites of injury.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag