Order Entry
France
ContactUsLinkComponent
Human recombinant PDGF R beta (from cells)
Human recombinant PDGF R beta (from cells)
Catalog # PRSI91-348
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant PDGF R beta (from cells)
Catalog # PRSI91-348
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    PDGFRB|Platelet-Derived Growth Factor Receptor 1|PDGFR|CD140 Antigen-like Family Member B|CD140b|Beta-Type Platelet-Derived Growth Factor Receptor|PDGFR1|PDGFR-Beta|Platelet-Derived Growth Factor Receptor Beta|Beta Platelet-Derived Growth Factor Receptor|PDGFR-1|PDGF-R-Beta
  • Protein/peptide name:
    PDGF R beta
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    57.17 kD
  • Sequence:
    Leu33-Phe530
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Platelet-Derived Growth Factor Receptor beta (PDGFR- beta ) is a member of the protein kinase superfamily and CSF-1/PDGF receptor subfamily. The PDGF family consists of PDGF-A, -B, -C and -D, which form either homo- or heterodimers (PDGF-AA, -AB, -BB, -CC, -DD). The four PDGFs are inactive in their monomeric forms. The PDGFs bind to the protein tyrosine kinase receptors PDGF receptor- alpha and - beta . These two receptor isoforms dimerize upon binding the PDGF dimer, leading to three possible receptor combinations, namely - alpha alpha, - beta beta and - alpha beta . The extracellular region of the PDGF receptor- beta consists of five immunoglobulin-like domains while the intracellular part is a tyrosine kinase domain. In addition to being a potent mitogen for cells of mesenchymal origin, PDGF has also been shown to be a potent chemoattractant for mesenchymal cells, mononuclear cells, and neutrophils and has been reported to be important in the modification of cellular matrix constituents.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag