- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:
- Environmentally Preferable:
- Biological activity:Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. The ED50 for this effect is typically 0. 5 - 8 ng/mL.
- Protein synonyms:SCF|KL-1|Kitl|KL|SF|FPH2|KITLG|MGF|SHEP7
- Protein/peptide name:SCF
- Purity:>98% as determined by SDS-PAGE.
- Molecular weight:19.1 kDa
- Formulation:Lyophilized, PBS, pH7.4
- Tested applications:Western Blot
Kit ligand (KITLG) is also known as stem cell factor (SCF), mast cell growth factor (MGF), steel factor (SF), which belongs to the SCF family, and is a widely expressed 28 - 40 kDa type I transmembrane glycoprotein. KITLG is the ligand for the receptor-type protein-tyrosine kinase KIT. SCF / MGF plays an essential role in the regulation of cell survival and proliferation, hematopoiesis, stem cell maintenance, gametogenesis, mast cell development, migration and function, and in melanogenesis. KITLG / SCF binding can activate several signaling pathways. KITLG / SF Promotes phosphorylation of PIK3R1, the regulatory subunit of phosphatidylinositol 3-kinase, and subsequent activation of the kinase AKT1. KITLG / SCF and KIT also transmit signals via GRB2 and activation of RAS, RAF1 and the MAP kinases MAPK1/ERK2 and/or MAPK3/ERK1. KITLG / SCF and KIT promote activation of STAT family members STAT1, STAT3 and STAT5. KITLG / SCF and KIT promote activation of PLCG1, leading to the production of the cellular signaling molecules diacylglycerol and inositol 1, 4, 5 - trisphosphate. KITLG / SCF acts synergistically with other cytokines, probably interleukins.