Order Entry
France
ContactUsLinkComponent
Human Recombinant Glutaredoxin 1 (from E. coli)
Human Recombinant Glutaredoxin 1 (from E. coli)
Catalog # PRSI96-355
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human Recombinant Glutaredoxin 1 (from E. coli)
Catalog # PRSI96-355
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Environmentally Preferable:
  • Protein synonyms:
    TTase-1|GRX1|GLRX|GRX|GLRX1
  • Protein/peptide name:
    Glutaredoxin 1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    12.7 kDa
  • Formulation:
    Lyophilized, 50 mM Tris, pH7.5 with 150 mM NaCl and 1 mM DTT
  • Tested applications:
    Western Blot

Specifications

About this item

Glutaredoxin-1 (GRX1) is also known as Thioltransferase-1 (TTase-1), GLRX, GRX, GLRX1, which belongs to the glutaredoxin family. GRX1 / GLRX contains one glutaredoxin domain, which exists in either a reduced or an oxidized form. GRX1 / GLRX has a glutathione-disulfide oxidoreductase activity in the presence of NADPH and glutathione reductase and functions as electron carriers in the glutathione-dependent synthesis of deoxyribonucleotides by the enzymeribonucleotide reductase.