Order Entry
France
ContactUsLinkComponent
 
Human recombinant CD84 (from HEK293 cells)
  PRSI96-109
 :  
undefined
Human recombinant CD84 (from HEK293 cells)
  PRSI96-109
 :  

 

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Environmentally Preferable:
  • Protein synonyms:
    LY9B|hCD84|mCD84|CD84|SLAMF5
  • Protein/peptide name:
    CD84
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    23.6 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

 

 

Leukocyte differentiation antigen CD84 is also known as SLAM family member 5 (SLAMF5), which belongs to immunoglobulin (Ig) superfamily. CD84 / SLAMF5 contains one Ig-like C2-type (immunoglobulin-like) domain. CD84 plays a role as adhesion receptor functioning by homophilic interactions and by clustering. CD84 / SLAMF5 increases proliferative responses of activated T-cells and SH2D1A/SAP does not seen be required for this process. Homophilic interactions enhance interferon gamma/IFNG secretion in lymphocytes and induce platelet stimulation via a SH2D1A/SAP-dependent pathway.