Order Entry
France
ContactUsLinkComponent
 
Human recombinant APOA-1 (from HEK293 cells)
Catalog # PRSI96-031
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant APOA-1 (from HEK293 cells)
Catalog # PRSI96-031
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Environmentally Preferable:
  • Biological activity:
    Immobilized Human ApoAI at 10μg/mL (100μl/well) can bind biotinylated human SCARB1. The EC50 of biotinylated human SCARB1 is 10-100 ng/mL.
  • Protein synonyms:
    Apolipoprotein A-I|MGC117399|APOA1
  • Protein/peptide name:
    APOA-1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    29 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

ApoA1 is also known as apolipoprotein A-I, ApoA-I , and is the major protein component of high density lipoprotein (HDL) in plasma. It has a specific role in lipid metabolism. Chylomicrons secreted from the intestinal enterocyte also contain ApoA1 but it is quickly transferred to HDL in the bloodstream. The protein promotes cholesterol efflux from tissues to the liver for excretion. It is a cofactor for lecithin cholesterol acyltransferase (LCAT) which is responsible for the formation of most plasma cholesteryl esters. ApoA-I was also isolated as a prostacyclin (PGI2) stabilizing factor, and thus may have an anticlotting effect. Defects in the gene encoding it are associated with HDL deficiencies, including Tangier disease, and with systemic non-neuropathic amyloidosis. In addition, it has been shown that ApoA1 is implicated in the anti-endotoxin function of HDL via interaction with lipopolysaccharide or endotoxin.