Human CMPK2 peptide (19 aa near C-terminus)
Catalog # PRSI7063P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Source:
- Species:Human
- Environmentally Preferable:
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:19 aa near C-terminus
- Protein/peptide name:CMPK2 Peptide (19 aa near C-terminus)
- Tested applications:Blocking
Specifications
About this item
CMPK2 peptide is used for blocking the activity of CMPK2 antibody.