Human TMEM18 peptide (15 aa near C-terminus)
Catalog # PRSI5043P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Source:
- Species:Human
- Environmentally Preferable:
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near C-terminus
- Protein/peptide name:TMEM18 Peptide (15 aa near C-terminus)
- Tested applications:Blocking
Specifications
About this item
TMEM18 peptide is used for blocking the activity of TMEM18 antibody.