- Antigen name:Tyrosine Hydroxylase
- Antigen symbol:TH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TH-4|tyrosine hydroxylase|EC 1.14.16.2|TYH|Tyrosine 3-hydroxylase|Tyrosine 3-monooxygenase|TY3H|TH isoform 3|TH isoform a
- Immunogen:TH polyclonal antibody was raised against native rat tyrosine hydroxylase, purified from pheochromocytoma.
- Size:100 µl
- Pk:100 µl
Tyrosine Hydroxylase (TH) is the rate-limiting enzyme in the synthesis of the catecholamines Dopamine and Norepinephrine. TH antibodies can therefore be used as markers for dopaminergic and noradrenergic neurons.
Tyrosine Hydroxylase antibody is specific for the 60k rat TH in lysates of PC-12 Cells. Applications include Immunofluorescence (IF), Immunohistochemistry (IHC) and Western Blot (WB). The antibody has been characterized in blot immunolabeling and immunocytochemical studies and cross-reacts with many different mammalian and some non-mammalian forms of TH. When internally tested under ideal conditions the working dilutions were 1:1000 for IHC, IF and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type:
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Bovine