Anti-Hepatitis C Mouse Monoclonal Antibody
PRSI35-519
- Antigen name:Hepatitis C
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Virus
- Western blot:Yes
- Form:Liquid
- Size:100 µg
- Pk:100 µG
Hepatitis C Virus antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.
Hepatitis C Virus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Hepatitis C
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse
Reactivity: Virus