Order Entry
France
ContactUsLinkComponent
 
Anti-OSBPL9 Rabbit Polyclonal Antibody
  PRSI30-091
undefined
Anti-OSBPL9 Rabbit Polyclonal Antibody
  PRSI30-091

 

  • Antigen name:
    Oxysterol binding Protein-like 9
  • Antigen symbol:
    OSBPL9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Dog
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    RP4-657D16.1|ORP9|ORP-9|oxysterol-binding protein-related protein 9|OSBP-related protein 9
  • Immunogen:
    A synthetic peptide corresponding a region of human OSBPL9.
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

OSBPL9 is a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain.This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. Multiple transcript variants, most of which encode distinct isoforms, have been identified.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: OSBPL9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human