Anti-EIF4E2 Rabbit Polyclonal Antibody
Catalog # PRSI29-341
Supplier: ProSci Inc.
Specifications
- Antigen symbol:EIF4E2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:mRNA cap-binding protein 4EHP|EIF4E2|mRNA cap-binding protein type 3|eIF-4E type 2|eIF4E-like protein 4E-LP|EIF4EL3|Eukaryotic translation initiation factor 4E type 2|Eukaryotic translation initiation factor 4E-like 3|eIF4E type 2|Eukaryotic translation initiation factor 4E homologous protein
- Immunogen:A synthetic peptide corresponding a region of human EIF4E2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
EIF4E2 belongs to the eukaryotic initiation factor 4E family. It recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EIF4E2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human