Specifications
- Antigen name:Transcription Factor 3
- Antigen symbol:TCF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:VDIR|E12|AW209082|A1|ME2|TCF-3|AA408400|Tcfe2a|bHLHb21|Pan2|E47|E2A|E12/E47|Pan1|ALF2
- Immunogen:A synthetic peptide corresponding a region of human TCF3.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
TCF3 contains 1 basic helix-loop-helix (bHLH) domain. Heterodimers between TCF3 and tissue-specific basic helix-loop-helix (bHLH) proteins play major roles in determining tissue-specific cell fate during embryogenesis, like muscle or early B-cell differentiation. Dimers bind DNA on E-box motifs: 5'-CANNTG-3'. TCF3 binds to the kappa-E2 site in the kappa immunoglobulin gene enhancer.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TCF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human