Anti-FOXO1 Rabbit Polyclonal Antibody
Catalog # PRSI28-788
Supplier: ProSci Inc.
Specifications
- Antigen name:Forkhead box protein O1
- Antigen symbol:FOXO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:FKHR|Forkhead in rhabdomyosarcoma|Forkhead box protein O1A|FOXO1
- Immunogen:A synthetic peptide corresponding a region of human FOXO1A.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
FOXO1A belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. The specific function of FOXO1A has not yet been determined; however, it may play a role in myogenic growth and differentiation. Translocation of FOXO1A gene with PAX3 has been associated with alveolar rhabdomyosarcoma.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FOXO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human