Order Entry
France
ContactUsLinkComponent
Anti-ZFP1 Rabbit Polyclonal Antibody
Anti-ZFP1 Rabbit Polyclonal Antibody
  PRSI27-370
undefined
Anti-ZFP1 Rabbit Polyclonal Antibody
  PRSI27-370

 

  • Antigen name:
    Zinc Finger Protein 1
  • Antigen symbol:
    ZFP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    FLJ34243|ZNF475
  • Immunogen:
    A synthetic peptide corresponding a region of human ZFP1.
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

ZFP1 is part of a large family of genes present in many organisms including yeast and human. Some of them are transcriptional activators and bind specifically to DNA by zinc mediated folded structures commonly known as zinc fingers.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ZFP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human