Order Entry
France
ContactUsLinkComponent
Anti-UPF1 Rabbit Polyclonal Antibody
Anti-UPF1 Rabbit Polyclonal Antibody
Catalog # PRSI25-352
Supplier:  ProSci Inc.
undefined
Anti-UPF1 Rabbit Polyclonal Antibody
Catalog # PRSI25-352
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    UPF1 Regulator of nonsense transcripts homolog
  • Antigen symbol:
    UPF1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    KIAA0221 |NORF1|EC 3.6.1.- |FLJ46894|pNORF1|FLJ43809 |HUPF1|hUpf1 |RENT1
  • Immunogen:
    A synthetic peptide corresponding a region of human UPF1.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

UPF1 is a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. UPF1 is located only in the cytoplasm. When translation ends, it interacts with the protein that is a functional homolog of yeast Upf2p to trigger mRNA decapping. Use of multiple polyadenylation sites has been noted for this gene.This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein is located only in the cytoplasm. When translation ends, it interacts with the protein that is a functional homolog of yeast Upf2p to trigger mRNA decapping. Use of multiple polyadenylation sites has been noted for this gene. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: UPF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human