Anti-BBX Rabbit Polyclonal Antibody
Catalog # PRSI25-120
Supplier: ProSci Inc.
Specifications
- Antigen name:Bobby Sox Homolog
- Antigen symbol:BBX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:MDS001|HBP2|HSPC339|ARTC1
- Immunogen:A synthetic peptide corresponding a region of human BBX.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
BBX is a transcription factor that is necessary for cell cycle progression from G1 to S phase.
BBX antibody can be used for detection of BBX by ELISA at 1:312500. BBX antibody can be used for detection of BBX by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: BBX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human