Specifications
- Antigen name:Transcription Factor CP2
- Antigen symbol:TFCP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human TFCP2.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
TFCP2 binds a variety of cellular and viral promoters including fibrinogen, alpha-globin, SV40 and HIV-1 promoters. Activation of the alpha-globin promoter in erythroid cells is via synergistic interaction with UBP1. It also functions as part of the SSP (stage selector protein) complex. TFCP2 facilitates the interaction of the gamma-globin genes with enhancer elements contained in the locus control region in fetal erythroid cells. It interacts by binding to the stage selector element (SSE) in the proximal gamma-globin promoter.
TFCP2 antibody can be used for detection of TFCP2 by ELISA at 1:312500. TFCP2 antibody can be used for detection of TFCP2 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TFCP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human