Carmustine, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Caution: Cold Shipment required
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Caution: Cold Shipment required
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1096724.
- 1096735.
- Description:
- Carmustine
- Carmustine Related Compound A
- Synonym(s):
- 1,3-Bis(2-chloroethyl)urea
- CAS No.:
- 154-93-8
- 2214-72-4
- Formula:
- C₅H₉Cl₂N₃O₂
- C₅H₁₀Cl₂N₂O
- UN No.:
- 2811
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1096724.CarmustineSpecifications:
More
Specifications
Cat. No.1096724.DescriptionCarmustineSynonym(s)CAS No.154-93-8FormulaC₅H₉Cl₂N₃O₂UN No.2811 - 1096735.Carmustine Related Compound ACarmustine Related Compound AUSP reference standardVial25 mgLog in for availability1 * 25 mg1 270,00 €Specifications:
More
Specifications
Cat. No.1096735.DescriptionCarmustine Related Compound ASynonym(s)1,3-Bis(2-chloroethyl)ureaCAS No.2214-72-4FormulaC₅H₁₀Cl₂N₂OUN No.



