Order Entry
France
Orders LinkContactUsLinkComponent
Sodium Picosulfate, Reference Standards

Sodium Picosulfate, Reference Standards

Supplier:   USP
Sodium Picosulfate, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1614578.
  • 1614589.
  • Description:
  • Sodium Picosulfate
  • Sodium Picosulfate Related Compound A
  • Synonym(s):
  • Sodium 4-((4-hydroxyphenyl)(pyridin-2-yl)methyl)phenyl sulfate
  • CAS No.:
  • 10040-45-6
  • 32500-19-9
  • Formula:
  • C₁₈H₁₃NO₈S₂.₂Na

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1614578.
    Sodium Picosulfate
    Sodium Picosulfate
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 408,00 €
    408,00 €
    1 * 50 mg
       408,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1614578.
    Description
    Sodium Picosulfate
    Synonym(s)
    CAS No.
    10040-45-6
    Formula
    C₁₈H₁₃NO₈S₂.₂Na
  • 1614589.
    Sodium Picosulfate Related Compound A
    Sodium Picosulfate Related Compound A
    USP reference standard
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 190,00 €
    1 190,00 €
    1 * 20 mg
       1 190,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1614589.
    Description
    Sodium Picosulfate Related Compound A
    Synonym(s)
    Sodium 4-((4-hydroxyphenyl)(pyridin-2-yl)methyl)phenyl sulfate
    CAS No.
    32500-19-9
    Formula
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.