Order Entry
France
Orders LinkContactUsLinkComponent
Risperidone, Reference Standards

Risperidone, Reference Standards

Supplier:   USP
Risperidone, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1604654.
  • 1604665.
  • 1604676.
  • 1604756.
  • Description:
  • Risperidone
  • Risperidone System Suitability Mixture
  • Risperidone Related Compounds Mixture
  • Risperidone Related Compound G
  • Synonym(s):
  • 3-{2-[4-(4-Fluoro-2-hydroxybenzoyl)piperidin-1-yl]ethyl}-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one hydrochloride
  • CAS No.:
  • 106266-06-2
  • 152542-00-2
  • Formula:
  • C₂₃H₂₇FN₄O₂
  • C₂₃H₂₈FN₃O₃
  • UN No.:
  • 2811
  • 2811
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1604654.
    Risperidone
    Risperidone
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 405,00 €
    405,00 €
    1 * 100 mg
       405,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1604654.
    Description
    Risperidone
    Synonym(s)
    CAS No.
    106266-06-2
    Formula
    C₂₃H₂₇FN₄O₂
    UN No.
    2811
  • 1604665.
    Risperidone System Suitability Mixture
    Risperidone System Suitability Mixture
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 1 280,00 €
    1 280,00 €
    1 * 10 mg
       1 280,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1604665.
    Description
    Risperidone System Suitability Mixture
    Synonym(s)
    CAS No.
    Formula
    UN No.
    2811
  • 1604676.
    Risperidone Related Compounds Mixture
    Risperidone Related Compounds Mixture
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 270,00 €
    1 270,00 €
    1 * 25 mg
       1 270,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1604676.
    Description
    Risperidone Related Compounds Mixture
    Synonym(s)
    CAS No.
    Formula
    UN No.
    2811
  • 1604756.
    Risperidone Related Compound G
    Risperidone Related Compound G
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 150,00 €
    1 150,00 €
    1 * 25 mg
       1 150,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1604756.
    Description
    Risperidone Related Compound G
    Synonym(s)
    3-{2-[4-(4-Fluoro-2-hydroxybenzoyl)piperidin-1-yl]ethyl}-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one hydrochloride
    CAS No.
    152542-00-2
    Formula
    C₂₃H₂₈FN₃O₃
    UN No.